Kinyoun's carbol fuchsin staining solution, for acid-fast bacteria

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Dimethyl fuchsin, Carbol-fuchsin, alpha4-(p-Aminophenyl)-alpha4-(4-imino-3-methyl-2,5-cyclohexadien-1-ylidene)-2,4-xylidine monohydrochloride

26087-06 26087-05
100504-482EA 219.72 USD
100504-482 100504-480
Kinyoun's carbol fuchsin staining solution, for acid-fast bacteria
Kinyoun's carbol fuchsin

Kinyoun's carbol fuchsin staining solution for acid-fast bacteria.


  • High quality staining solution
  • Ready-to-use

Formula: C₂₁H₂₂ClN₃
MW: 351.878 g/mol
Boiling Pt: 100 °C (212 °F)
Storage Temperature: Ambient
CAS Number: 26261-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR