2-Propanol ACS

Supplier: Electron Microscopy Sciences
Danger

16931 16930
100504-064EA 109.72 USD
100504-064 100504-062
2-Propanol ACS
2-Propanol

Isopropyl Alcohol, Reagent A.C.S.: (2-Propanol).

Formula: (CH₃)₂CHOH
Boiling Pt: 82 °C (180 °F)
Melting Pt: –89.5 °C (–129 °F)
Density: 0.785 g/cm³ (6.551 lbs/gal.)
Flash Pt: 13 °C (55 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8249b13ac2b2348367ff2a74adc1fd5b2fdd24e3&vid=8249b13ac2b2348367ff2a74adc1fd5b2fdd24e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10
Titratable acid or Base 0.0001 meq/g
Residue after evaporation 0.001%
Solubility in water To pass test
Water 0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR