Hexamethylenetetramine 99-100.5%, granules ACS
Supplier: Electron Microscopy Sciences
Synonyms:
Hexamethylenetetramine, Hexamine, Urotropine, Formin
16710
16710-25
100504-034EA
28.19
USD
100504-034
100502-404
Hexamethylenetetramine 99-100.5%, granules ACS
Hexamethylenetetramine
(Urotropine; Methenamine)
C6N4 F.W. 140.19
Assay: 99.0-100.5%
Used in silver stains.
Swift, J.A. (1968) Microsc. Soc. 88, 449.
Used in silver stains.
Formula:
C₆H₁₂N₄ MW: 140.19 g/mol |
MDL Number:
MFCD00006895 CAS Number: 100-97-0 UN: 1328 ADR: 4.1,III |
Specification Test Results
Assay: | 99.0-100.5% |
Residue after Ignition | 0.1% |
Chloride | 0.014% |
Sulfate | To Pass Test |
Heavy Metals. | 0.001% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...