HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99%, crystals biological buffer
Supplier: Electron Microscopy Sciences
Synonyms:
HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
16780
16782
100504-040EA
27.16
USD
100504-040
100504-042
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99%, crystals biological buffer
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid)
Formula:
C₈H₁₈N₂O₄S MW: 238.31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006158 CAS Number: 7365-45-9 |
Specification Test Results
Assay | > 99% |
pKa @20°C | 7.55 |
pH | 7.0-8.0 |
A(260) (1.0M in H2O) | <0.05 |
Ash | <0.1% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...