Gram's iodine solution
Supplier: Electron Microscopy Sciences
|
Warning
|
Synonyms:
Lugol's iodine, Iodine and potassium iodide, Potassium triiodide
26054-06
26054-05
100504-438EA
68.93
USD
100504-438
100504-436
Gram's iodine solution
Gram's iodine solution
For use as a bacterial, fungal and inclusion body stain.
- High quality staining solution
- Ready-to-use
Formula:
I₃K MW: 419.812 g/mol Boiling Pt: 100 °C (212 °F) Density: 1.03891 g/cm³ (8.67 lbs/gal.) Storage Temperature: Ambient |
MDL Number:
MFCD00134128 CAS Number: 12298-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...