tetra-Chloroauric(III) acid trihydrate ≥99.998%

Supplier: Electron Microscopy Sciences
Danger

16584 16582 16583
100504-800BX 2635.9 USD
100504-800 100504-798 100504-944
tetra-Chloroauric(III) acid trihydrate ≥99.998%
tetra-Chloroauric(III) acid trihydrate

A highly purified gold compound for the preparation of colloidal gold sols. Preweighed to facilitate your work.


Información de suministro: It includes preparation instructions for gold sols.

Formula: HAuCl₄·3H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149904
CAS Number: 16961-25-4
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 99.9985%
Gold contents 49%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR