Giemsa's stain solution, certified by the Biological Stain Commission
Supplier: Electron Microscopy Sciences
|
Danger
|
Synonyms:
Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue
Used to differentiate the types of leucocytes, bacteria and inclusion bodies (Giemsa, Centralbl. F. Publ., Abt. 1,37, 308 (1904)), stain for thick blood films; shows malarial parasites (Barber and Komp, Publ. Health Rep., 44, 2330 (1929)), bone marrow and stain for showing cell morphology; together with May-Grunwald Solution for pap staining (Pappenheim, Folia Haemat., 13, 338 (1912)).
It contains glycerin (61%), methyl alcohol (38.3%), azur II (0.16%), azur II eosin (0.6%).
Boiling Pt:
64 °C (147 °F) Flash Pt: 11 °C (52 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00081642 CAS Number: 51811-82-6 UN: 1230 ADR: 3,II |
Specification Test Results
Specific Gravity | 0.89 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...