Eosin Y (yellowish), certified (C.I. 45380)
Supplier: Electron Microscopy Sciences
|
Warning
|
14850
14851
100503-872EA
21.9
USD
100503-872
100503-874
Eosin Y (yellowish), certified (C.I. 45380)
Eosin Y (yellowish)
Eosin Y can be used with hematoxylin for showing cell nuclei in embryos and kidneys.
- Component of Wright's stain for blood corpuscles
- For staining bone marrow to show cell morpholog
Solubility : 44.0% Water; 2.0% Alcohol; 25.0% Cellosolve; 27.5% Glycol; 0.0% Xylene
Formula:
C₂₀H₆Br₄Na₂O₅ Density: 1.018 g/cm³ (8.495 lbs/gal.) Storage Temperature: Ambient |
MDL Number:
MFCD00005040 CAS Number: 17372-87-1 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...