D.E.R.™ 736 Resin
Supplier: Electron Microscopy Sciences
|
Warning
|
13001
13000
100503-794EA
327.28
USD
100503-794
100503-792
D.E.R.™ 736 Resin
D.E.R.™ 736 Resin
A light colored, flexible epoxy.
Used to simplify infiltration in combination with Epon 812 (Embed 812), Kushida, J. Electron Micro., 16,278 (1964).
Storage Temperature:
Ambient |
CAS Number:
41638-13-5 |
Specification Test Results
Epoxide Equivalent Weight | 175 - 205 |
Specific Gravity | 1.14 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...