Dodecenylsuccinic anhydride (mixture of isomers), specially distilled
Supplier: Electron Microscopy Sciences
|
Warning
|
DDSA (Dodenyl Succinic Anhydride) specially distilled is an ultra pure DDSA that is distilled by a new, slower process which has a distinct effect on the color control of embedding resins and offers complete infiltration of tissues. Produces blocks which are totally transparent.
Formula:
C₁₆H₂₆O₃ Boiling Pt: 350 °C (662 °F) Density: 9.2 g/cm³ (76.774 lbs/gal.) Flash Pt: 167 °C (333 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00036197 CAS Number: 25377-73-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...