Dodecenylsuccinic anhydride (mixture of isomers)

Supplier: Electron Microscopy Sciences
Warning

13701 13700
100503-842EA 111.34 USD
100503-842 100503-840
Dodecenylsuccinic anhydride (mixture of isomers)
Dodecenylsuccinic anhydride (mixture of isomers)

(Dodecenyl Succinic Anhydride) C16H26O3 F.W. 266.38 CAS 2654-438-7 Specific Gravity: 1.005

Formula: C₁₆H₂₆O₃
Boiling Pt: 182 °C (360 °F)
Melting Pt: 43 °C (109 °F)
Density: 1.007 g/cm³ (8.403 lbs/gal.)
Flash Pt: 178 °C (352 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036197
CAS Number: 25377-73-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=371366918112b469dab2f249f6c32546e8ee5677&vid=371366918112b469dab2f249f6c32546e8ee5677&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Specific Gravity 1.005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR