Chloroform ACS

Supplier: Electron Microscopy Sciences

12541 12540
100504-934EA 157.2 USD
100504-934 100503-758
Chloroform ACS
Chloroform

CHCl3 FW 119.38 CAS 67-66-3 A.C.S.

Formula: CHCl₃
MDL Number: MFCD00000826
CAS Number: 67-66-3
UN: 1888
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd7b089b86eb10623b4c77ccca3213cbc0d47c83&vid=f01efdcdf34b24309a77803e09d1562bc60047ba&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR