Cadmium(II) iodide ≥99.0%, crystals ACS

Supplier: Electron Microscopy Sciences
Danger

12321 12320
100503-750EA 265.01 USD
100503-750 100503-748
Cadmium(II) iodide ≥99.0%, crystals ACS
Cadmium(II) iodide

Used for negative staining. Possible Carcinogen.

Formula: CdI₂
Boiling Pt: 796 °C (1465 °F)
Melting Pt: 387 °C (729 °F)
Density: 5.67 g/cm³ (47.316 lbs/gal.)
MDL Number: MFCD00010920
CAS Number: 7790-80-9
UN: 2570
ADR: 6.1,III

Order Now

Specification Test Results

Assay Min. 99.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR