Cacodylic acid (dimethylarsinic acid) free acid ≥99.5%

Supplier: Electron Microscopy Sciences
Danger

12200 12201
100503-742EA 133.87 USD
100503-742 100503-744
Cacodylic acid (dimethylarsinic acid) free acid ≥99.5%
Cacodylic acid (dimethylarsinic acid)

(Dimethylarsinic Acid) (CH3)2AsO2H F.W. 137.99 CAS 75-60-5 Assay 99.5%

Formula: (CH₃)₂As(O)OH
MDL Number: MFCD00002095
CAS Number: 75-60-5
UN: 1572
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6337f62c3acf3a32aff5eac7b9e31cc77cac8c1&vid=0046383924a44615a807695749099f01cb069b00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR