Bromophenol blue ACS

Supplier: Electron Microscopy Sciences

11471 11470
100503-700EA 151.59 USD
100503-700 100503-698
Bromophenol blue ACS
Bromophenol blue

(3',3",5 '5"-Tetrabromophenol-sulfonthalein) C19H10O5SBr4 F.W. 669.97 CAS 115-39-9

Formula: C₁₉H₁₀Br₄O₅S
Density: 1.3 g/cm³ (10.849 lbs/gal.)
MDL Number: MFCD00005875
CAS Number: 115-39-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68d102ce97a6648b0581974e2760784338527e81&vid=68d102ce97a6648b0581974e2760784338527e81&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR