Amyl acetate, purified, Glass Distilled
Supplier: Electron Microscopy Sciences
|
Warning
|
10816
10815
100503-662EA
167.12
USD
100503-662
100503-660
Amyl acetate, purified, Glass Distilled
Amyl acetate
Purified, glass distilled amyl acetate is available to meet your laboratory needs.
Formula:
C₇H₁₄O₂ Boiling Pt: 149 °C (300 °F) Density: 0.88 g/cm³ (7.344 lbs/gal.) Flash Pt: 38 °C (100 °F) |
MDL Number:
MFCD00009500 CAS Number: 628-63-7 UN: 1104 ADR: 3,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...