Agar, Powder, Bacteriological Grade, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
10202
10200
100503-640EA
430.79
USD
100503-640
100505-012
Agar, Powder, Bacteriological Grade, Electron Microscopy Science
Media for Microbiology
Dehydrated Media for Microbiology
Agar, Powder has a gelling temperature (1.5% in H2O) ~35 °C
Gel Strength (1.5% gel) 300g/cm2.