Acetone, Glass Distilled

Supplier: Electron Microscopy Sciences
Danger

10016 10015
100503-624EA 139.26 USD
100503-624 100503-622
Acetone, Glass Distilled
Acetone

U.V. Absorption - 1cm cell vs. H2O.

Formula: CH₃COCH₃
Boiling Pt: 56.5 °C (134 °F)
Melting Pt: 94.7 °C (202 °F)
Density: 0.79 g/cm³
Flash Pt: –20 °C (–4 °F)
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

U.V. Absorption (1 cm cell vs. H₂O) 0.01, 0.02, 0.10, and 1.0 at Wavelengths of 400, 350, 340, and 330 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR