Toluene-α-sulfonyl fluoride ≥99.0%

Supplier: Electron Microscopy Sciences
Danger

Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulphonyl fluoride

19330 19331
100504-188EA 49.55 USD
100504-188 100504-190
Toluene-α-sulfonyl fluoride ≥99.0%
Toluene-α-sulphonyl fluoride

C6H5CH2SO2F F.W. 174.20 CAS 329-98-6

Formula: C₇H₇FO₂S
MW: 174.2 g/mol
Melting Pt: 92…94 °C (198…201 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007424
CAS Number: 329-98-6
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfb3334a86d56c4d75cfd417f5304acfa785cf0e&vid=dfb3334a86d56c4d75cfd417f5304acfa785cf0e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay >99.0%
Water <0.1%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR