2-Hydroxypropyl methacrylate (mixture of isomers) inhibited

Supplier: Electron Microscopy Sciences

Synonyms: Propylene Glycol Monomethacrylate, HPMA, Hydroxypropyl methacrylate, Methacrylic acid 2-hydroxypropyl ester, Methacrylic acid hydroxypropyl ester

Stabilization: ∼300 ppm Stabilised with 4-methoxyphenol (MEHQ)

16900 16901
100504-050EA 54.59 USD
100504-050 100504-946
2-Hydroxypropyl methacrylate (mixture of isomers) inhibited
2-Hydroxypropyl methacrylate (mixture of isomers)

2-Hydroxypropyl Methacrylate is a water miscible monomer. Inhibited with approximately 300ppm MEHQ.


Solubility in water 13%

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 70 °C (1 mmHg)
Density: 1.026 g/cm³ (8.562 lbs/gal.)
Flash Pt: 98 °C (208 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004536
CAS Number: 27813-02-1
UN: 1919
ADR: 3.2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0536ef7a39424aa10c33f12a2a8263c4196e424d&vid=0536ef7a39424aa10c33f12a2a8263c4196e424d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Specific Gravity @25 °C 1.028


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR