Benzyl(dimethyl)amine ≥98%

Supplier: Electron Microscopy Sciences
Danger

Synonyms: DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA

11400-25 11400
100496-392EA 26.02 USD
100496-392 100503-690
Benzyl(dimethyl)amine ≥98%
Benzyl(dimethyl)amine

An amine accelerator for polymerization of epoxy resins. A direct alternative to DMP-30 with a longer shelf life and better penetration.

Formula: C₆H₅CH₂N(CH₃)₂
MW: 135.21 g/mol
Boiling Pt: 184 °C (363.2 °F)
Melting Pt: –75 °C (–103 °F)
Density: 0.9 g/cm³ (7.5105 lbs/gal.)
Flash Pt: 54 °C (129.2 °F)
MDL Number: MFCD00008329
CAS Number: 103-83-3
UN: 2619
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f02d317b7c79caba024b45c17e92be61101aa0fd&vid=f02d317b7c79caba024b45c17e92be61101aa0fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR