Schroeder Tenaculum Forceps, OR Grade, Sklar
Supplier: Sklar
91-1298 91-1290
10192-354EA 172.97 USD
10192-354 10193-524
Schroeder Tenaculum Forceps, OR Grade, Sklar
Forceps
Schroeder-Braun Tenaculum Forceps are a ratcheted, finger ring instrument used in OB/GYN procedures involving manipulation of the cervix or uterus, D&C, and vaginal or abdominal hysterectomy
  • The tips of the Schroeder-Pozzi tenaculum forceps have two rounded, inward curving prongs and are traumatic to tissue
  • The tips of the Schroeder-Braun tenaculum forceps have two flattened prong tips that angle inward at 90 degrees
  • The prongs overlap, providing a strong hold on the tissue, but are also traumatic
  • They have straight shanks and an overall length of 10 inches
They are also used to grasp, hold, and move vessels and arteries during surgical procedures. The tips of the Schroeder-Braun tenaculum forceps have two flattened prong tips that angle inward at 90 degrees. The prongs overlap, providing a strong hold on the tissue, but are also traumatic.
Información para pedidos: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Precaución: Customers must have a Censitrac Site ID to purchase Censitrac instruments.