Merit™ Rochester-Ochsner Forceps, Physician Grade, Sklar
Supplier: Sklar
97-500 97-501 97-504 97-513 97-515 97-517 97-521 97-512
10184-836EA 23.17 USD
10184-836 10184-838 10184-840 10184-844 10184-846 10184-848 10184-850 10193-868
Merit™ Rochester-Ochsner Forceps, Physician Grade, Sklar
Forceps
Merit™ Rochester-Oschner (Kocher) Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels.
They have horizontal serrations the entire length of the jaw as well as 1 x 2 teeth at the tip. The combination of full serrations plus teeth ensure a firm grip on the tissue or vessel being held. Rochester-Oschner Forceps are available either curved or straight and in multiple lengths. These forceps are sometimes referred to as simply Oschner Forceps, however, Rochester-Oschner Forceps are actually heavier than standard Oschner Forceps.
Merit™ instruments are mid-grade products used for routine and in-office procedures. These high quality instruments are crafted from Pakistan steel with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices that require quality, durable instruments.