Rochester-Pean Forceps, OR Grade, Sklar
Supplier: Sklar
17-2267 17-2362 17-2467 22-2272
10192-448EA 105.6 USD
10192-448 10194-756 10194-758 10195-210
Rochester-Pean Forceps, OR Grade, Sklar
Forceps
Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. Available curved or straight. The forceps are constructed with stainless steels. Censitrac Ready OR Grade Rochester-Pean Forceps are industry standard.
Información para pedidos: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Precaución: Customers must have a Censitrac Site ID to purchase Censitrac instruments.