Human Recombinant GRO-beta (from E. coli)
Supplier: Peprotech
300-39-2UG
300-39-10UG
300-39-100UG
10780-538EA
143.88
USD
10780-538
10780-540
10780-542
Human Recombinant GRO-beta (from E. coli)
Proteins and Peptides
All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils. Recombinant Human GRO-β (CXCL2) is a 7.9 kDa protein consisting of 73 amino acids, including the 'ELR' motif common to the CXC chemokine family that binds to CXCR1 or CXCR2.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.