Mouse Recombinant I-TAC (from E. coli)
Supplier: Peprotech
250-29-5UG
250-29-20UG
250-29-100UG
250-29-250UG
250-29-500UG
250-29-1MG
10780-312EA
132.81
USD
10780-312
10780-314
10780-316
10780-318
10780-320
10780-322
Mouse Recombinant I-TAC (from E. coli)
Proteins and Peptides
I-TAC is a "non-ELR" CXC chemokine that is regulated by interferon and signals through the CXCR3 receptor. I-TAC is chemoattractant for IL-2-activated T cells, but does not affect freshly isolated un-stimulated T cells, neutrophils, or monocytes. Recombinant Murine I-TAC (CXCL11) is a 9.0 kDa protein containing 79 amino acid residues, including the four highly conserved cysteine residues present in CXC chemokines.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.