Mouse Recombinant IL-9 (from E. coli)
Supplier: Peprotech
219-19-2UG 219-19-10UG 219-19-50UG 219-19-100UG 219-19-250UG 219-19-500UG 219-19-1MG
10780-196EA 143.41 USD
10780-196 10780-198 10780-200 10780-202 10780-204 10780-206 10780-208
Mouse Recombinant IL-9 (from E. coli)
Proteins and Peptides
IL-9 is an immunoregulatory cytokine produced by IL-2 activated Th2 lymphocytes. IL-9 enhances the proliferation of T lymphocytes, mast cells, erythroid precursor cells and megakaryoblastic leukemia cell lines. Over-expression of IL-9 has been implicated in the pathogenesis of anaplastic lymphoma and Hodgkin’s disease. Whereas murine IL-9 can function on human cells, human IL-9 is inactive on mouse cells. Recombinant Murine IL-9 is a 14.3 kDa protein of 127 amino acid residues, including 10 cysteine residues that are fully conserved between the human murine proteins.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.