Human Recombinant Gremlin-1 (from CHO cells)
Supplier: Peprotech
120-42-10UG 120-42-50UG 120-42-100UG 120-42-250UG 120-42-500UG 120-42-1MG
10779-250EA 143.88 USD
10779-250 10779-252 10779-254 10779-256 10779-258 10779-260
Human Recombinant Gremlin-1 (from CHO cells)
Proteins and Peptides
Gremlin-1 (isoform-1) belongs to a group of diffusible proteins that bind to ligands of the TGF-β family and regulate their activity by inhibiting their access to signaling receptors. The interplay between TGF-β ligands and their natural antagonists has major biological significance during development processes, in which cellular response can vary considerably depending upon the local concentration of the signaling molecule. Gremlin-1 is highly expressed in the small intestine, fetal brain, and colon; and is expressed at lower levels in the brain, prostate, pancreas, and in skeletal muscle. Gremlin-1 regulates multiple functions in early development by specifically binding to, and inhibiting the function of, BMP-2, -4, and -7. It also plays a role in carcinogenesis and kidney branching morphogenesis. Recombinant Human Gremlin-1 is an 18.3 kDa protein containing 160 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.