Human Recombinant NP-1 (from E. coli)
Supplier: Peprotech
300-42-5UG 300-42-20UG 300-42-50UG 300-42-100UG 300-42-250UG 300-42-500UG 300-42-1MG
10773-952EA 132.81 USD
10773-952 10773-954 10773-956 10773-958 10773-960 10773-962 10773-964
Human Recombinant NP-1 (from E. coli)
Proteins and Peptides
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The α-defensins, which include NP-1, NP-2 and NP-3, are distinguished from the β-defensins by the pairing of their three disulfide bonds. In addition to antimicrobial activity, NP-1 exhibits chemotactic activity on dendritic cells. NP-1 is expressed as the C-terminal portion of an inactive precursor protein, which also contains a 19 amino acid N-terminal signal sequence and a 45 amino acid polypeptide. NP-1 contains a six-cysteine motif that forms three intra-molecular disulfide bonds. Recombinant Human NP-1 is a 3.4 kDa protein containing 30 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.