Human Recombinant GRO-beta (from E. coli)
Supplier: Peprotech
300-39-250UG 300-39-500UG 300-39-1MG
10773-922EA 1615.45 USD
10773-922 10773-924 10773-926
Human Recombinant GRO-beta (from E. coli)
Proteins and Peptides
All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils. Recombinant Human GRO-β (CXCL2) is a 7.9 kDa protein consisting of 73 amino acids, including the 'ELR' motif common to the CXC chemokine family that binds to CXCR1 or CXCR2.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.