Human Recombinant MCP-4 (from E. coli)
Supplier: Peprotech
300-24-5UG 300-24-20UG 300-24-100UG 300-24-250UG 300-24-500UG 300-24-1MG
10773-692EA 132.81 USD
10773-692 10773-694 10773-696 10773-698 10773-700 10773-702
Human Recombinant MCP-4 (from E. coli)
Proteins and Peptides
The MCP proteins are members of the CC chemokine family that signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross-reacts across species. Recombinant Human MCP-4 (CCL13) is an 8.6 kDa protein containing 75 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.