Human Recombinant ENA-78 (from E. coli)
Supplier: Peprotech
ENA-78 is a CXC chemokine that signals through the CXCR2 receptor. It is expressed in monocytes, platelets, endothelial cells, and mast cells. ENA-78 is a chemoattractant for neutrophils. Three N-terminal truncated variants of human ENA-78; ENA 5-78, ENA 8-78, ENA 9-78, contain 74, 71, and 70 amino acid residues, respectively, possess increased biological activity. ENA-78 contains the four conserved cysteine residues present in CXC chemokines, and also contains the ‘ELR’ motif common to CXC chemokine that bind to the CXCR1 and CXCR2 receptors. Recombinant Human ENA-78 (CXCL5) is a 7.8 kDa protein consisting of 71 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.