Human Recombinant ENA-78 (from E. coli)
Supplier: Peprotech
300-22-5UG 300-22-20UG 300-22-50UG 300-22-100UG 300-22-250UG 300-22-500UG 300-22-1MG
10773-652EA 132.81 USD
10773-652 10773-654 10773-656 10773-658 10773-660 10773-662 10773-664
Human Recombinant ENA-78 (from E. coli)
Proteins and Peptides
ENA-78 is a CXC chemokine that signals through the CXCR2 receptor. It is expressed in monocytes, platelets, endothelial cells, and mast cells. ENA-78 is a chemoattractant for neutrophils. The three naturally occurring variants of human ENA-78; ENA 5-78, ENA 9-78 and ENA 10-78, contain 74, 70, and 69 amino acid residues, respectively, and possess the same biological activity. ENA-78 contains the four conserved cysteine residues present in CXC chemokines, and also contains the ‘ELR’ motif common to CXC chemokine that bind to the CXCR1 and CXCR2 receptors. Recombinant Human ENA-78 (CXCL5) is an 8.0 kDa protein consisting of 74 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.