Human Recombinant GRO-alpha/MGSA (from E. coli)
Supplier: Peprotech
300-11-5UG
300-11-25UG
300-11-100UG
300-11-250UG
300-11-500UG
300-11-1MG
10773-542EA
141.62
USD
10773-542
10773-544
10773-546
10773-548
10773-550
10773-552
Human Recombinant GRO-alpha/MGSA (from E. coli)
Proteins and Peptides
All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils. GRO/MGSA also stimulates mitogenesis in certain human melanoma cells. Recombinant Human GRO-α/MGSA (CXCL1) is a 7.8 kDa protein consisting of 73 amino acids, including the 'ELR' motif common to the CXC chemokine family that binds to CXCR1 or CXCR2.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.