Mouse Recombinant C10 (from E. coli)
Supplier: Peprotech
250-06-2UG 250-06-10UG 250-06-100UG 250-06-250UG 250-06-500UG 250-06-1MG
10773-160EA 139.89 USD
10773-160 10773-162 10773-164 10773-166 10773-168 10773-170
Mouse Recombinant C10 (from E. coli)
Proteins and Peptides
Murine C10 belongs to the CC chemokine family and is expressed in myelopoietic bone marrow cultures when stimulated with GM-CSF, M-CSF, IL-3 or IL-4. It signals primarily through the CCR1 receptor. C10 is chemotactic for B cells, CD4+ T cells, monocytes and NK cells and also exhibits powerful suppressive activity on colony formation by different lineages of hematopoietic progenitors. The C10 contains the four highly conserved cysteine residues present in CC chemokines. The mature protein contains 95 amino acid residues. Recombinant Murine C10 (CCL6) is a 10.7 kDa protein containing 95 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.