Human Recombinant GLP-1 (from E. coli)
Supplier: Peprotech
130-08-200UG 130-08-1MG 130-08-5MG
10772-722EA 123.33 USD
10772-722 10772-724 10772-726
Human Recombinant GLP-1 (from E. coli)
Proteins and Peptides
GLP-1 is a proglucagon-derived peptide hormone secreted primarily by intestinal L cells during feeding. Its major physiological function is the stimulation of pancreatic β cells to release appropriate amounts of insulin after glucose absorption. Other biological actions exhibited by GLP-1 include the suppression of plasma glucagon levels, inhibition of gastric motility, and promotion of satiety. The secretion of GLP-1 from intestinal L cells is stimulated by nutrients, hormones, and neural inputs. On the other hand, insulin has been reported to inhibit GLP-1 release, indicating that a feedback loop mechanism regulates GLP-1 secretion. In addition to being the precursor of GLP-1, proglucagon, whose primary structure is highly conserved in mammalian species, is also the precursor for other members of the glucagon family of peptide hormones, including glicentin-related pancreatic peptide (GRPP), glucagons, and GLP-2. Recombinant Human GLP-1 is a 3.3 kDa polypeptide consisting of 31 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.