Human Recombinant PlGF-3 (from E. coli)
Supplier: Peprotech
100-57-5UG
100-57-25UG
100-57-100UG
100-57-250UG
100-57-500UG
100-57-1MG
10772-294EA
132.81
USD
10772-294
10772-296
10772-298
10772-300
10772-302
10772-304
Human Recombinant PlGF-3 (from E. coli)
Proteins and Peptides
PlGF-3 is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-3 is expressed exclusively in the placenta. It signals through the VEGFR-1/FLT1 receptor, and stimulates endothelial cell proliferation and migration. PlGF-3 lacks heparin binding affinity. Recombinant Human PlGF-3 is a 45.7 kDa disulfide-linked homodimeric protein of two 203 amino acid polypeptide chains.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.