Cast Iron Support Rings
Supplier: United Scientific Supplies
SRCI05 SRCI06 SRCI07 SRCI02 SRCI03 SRCI04
470157-178EA 14.4 USD
470157-178 470157-180 470157-182 470157-266 470157-268 470157-270
Cast Iron Support Rings
Support Rings
These zinc-plated support rings are made of durable cast iron.
  • Zinc-Plated
  • Cast Iron Construction
  • Boss-Head Clamp and Thumbscrew Included
  • Several Diameters Available
  • Accommodates rods up to ¾”
They're equipped with a boss-head clamp and thumbscrew.
Support rings are available in several diameters.