Merit™ Metzenbaum Scissors, Physician Grade, Sklar
Supplier: Sklar
97-335 97-339 97-340 97-336
10184-410EA 22.05 USD
10184-410 10184-412 10184-414 10193-658
Merit™ Metzenbaum Scissors, Physician Grade, Sklar
Scissors
Merit™ Metzenbaum dissecting scissors are used for dissecting and cutting fine tissue.
Metzenbaum scissors are identifiable by their long, thin shanks and short jaws with fine, blunt, rounded tips. These scissors are not recommended for cutting sutures, drains, or heavy tissue.
Merit™ instruments are office-grade line of products used for routine and in-office minor procedures. These high quality instruments are crafted from Pakistan steel and come with a ten year guarantee. Merit™ instruments deliver a solution for physicians and practices, that require quality and durable instruments.