3-Amino-6-bromo-4-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
5-Amino-2-bromo-4-methylpyridine, 98%
Purity: 98%
CAS No.: 156118-16-0
Molecular Formula: C6H7BrN2
Formula Weight: 187.04
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Brown
Beilstein Reference: 10654630
UN No.: 2811
MDL No.: MFCD08277280
Formula:
C₆H₇BrN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD08277280 CAS Number: 156118-16-0 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...