1-Bromo-3,5-dinitrobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,5-Dinitrobromobenzene
H66774-03
H66774-06
AAH66774-03EA
48.9
USD
AAH66774-03
AAH66774-06
1-Bromo-3,5-dinitrobenzene 98%
1-Bromo-3,5-dinitrobenzene
1-Bromo-3,5-dinitrobenzene, 98%
Purity: 98%
CAS No.: 18242-39-2
Molecular Formula: C6H3BrN2O4
Formula Weight: 247.00
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 2053146
UN No.: 3459
MDL No.: MFCD00156596
Formula:
C₆H₃BrN₂O₄ MW: 247 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00156596 CAS Number: 18242-39-2 UN: 3459 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...