3-Amino-2,4-dimethylpyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
3-Amino-2,4-dimethylpyridine, 97%
Purity: 97%
CAS No.: 1073-21-8
Molecular Formula: C7H10N2
Formula Weight: 122.17
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: Pale brown
Beilstein Reference: 111999
UN No.: 2811
MDL No.: MFCD08235192
Formula:
C₇H₁₀N₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD08235192 CAS Number: 1073-21-8 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...