α-Bromo-4-fluorophenylacetic acid 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Bromo-2-(4-fluorophenyl)acetic acid
H66692-03
H66692-06
AAH66692-03EA
59.9
USD
AAH66692-03
AAH66692-06
α-Bromo-4-fluorophenylacetic acid 96%
α-Bromo-4-fluorophenylacetic acid
^a-Bromo-4-fluorophenylacetic acid, 96%
Purity: 96%
CAS No.: 29270-33-5
Molecular Formula: C8H6BrFO2
Formula Weight: 233.03
Physical Form: Powder
Appearance: White to brown
Melting Point: 96-100°
UN No.: 3261
MDL No.: MFCD08276759
Formula:
C₈H₆BrFO₂ MW: 233.03 g/mol Melting Pt: 96…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD08276759 CAS Number: 29270-33-5 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...