Pyruvonitrile 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
H66636-03
H66636-06
AAH66636-03EA
49.8
USD
AAH66636-03
AAH66636-06
Pyruvonitrile 90%, Technical Grade
Pyruvonitrile
Pyruvonitrile, tech. 90%
Purity: 90%
CAS No.: 631-57-2
Molecular Formula: C3H3NO
Formula Weight: 69.06
Storage Temperature: 2°C to 8°C
Flash Point: 14°(57°F)
Physical Form: Liquid
Density: 0.974
Refractive Index: 1.3755
Appearance: Colorless to brown
Boiling Point: 92-93°
Beilstein Reference: 1737633
UN No.: 3273
MDL No.: MFCD00001838
Formula:
C₃H₃NO MW: 69.063 g/mol Boiling Pt: 92…93 °C Density: 0.974 Flash Pt: 14 °C (57 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001838 CAS Number: 631-57-2 EINECS: 211-159-2 UN: 3273 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...