(S)-(+)-1-Boc-3-(methylamino)piperidine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
(S)-(+)-1-Boc-3-(methylamino)piperidine, 96%
Purity: 96%
CAS No.: 912368-73-1
Molecular Formula: C11H22N2O2
Formula Weight: 214.30
Storage Temperature: 2°C to 8°C
Flash Point: 110°(230°F)
Physical Form: Liquid
Density: 0.985
Refractive Index: 1.4600
Sensitivity: Moisture Sensitive
Appearance: Colorless to yellow
Optical Rotation: +6.0° (c=0.5 in chloroform)
UN No.: 2810
MDL No.: MFCD09752964
Formula:
C₁₁H₂₂N₂O₂ Density: 0.985 Flash Pt: 110 °C(230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD09752964 CAS Number: 912368-73-1 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...