Sodium-4-(tert-butylcarbonyloxy)benzenesulfonate 95%

Supplier: Thermo Scientific Chemicals
Warning

H66496-06 H66496-14
AAH66496-03EA 23.17 USD
AAH66496-03 AAH66496-06 AAH66496-14
Sodium-4-(tert-butylcarbonyloxy)benzenesulfonate 95%
Sodium-4-(tert-butylcarbonyloxy)benzenesulfonate

Sodium 4-(tert-butylcarbonyloxy)benzenesulfonate, 95%


Purity: 95% CAS No.: 188114-91-2 Molecular Formula: C11H13NaO5S Formula Weight: 280.27 Physical Form: Solid MDL No.: MFCD09859793

Formula: C₁₁H₁₃NaO₅S
MW: 280.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09859793
CAS Number: 188114-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR