2-Amino-6-methoxypyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66436-06
H66436-14
AAH66436-06EA
74.1
USD
AAH66436-06
AAH66436-14
2-Amino-6-methoxypyridine 97%
2-Amino-6-methoxypyridine
2-Amino-6-methoxypyridine, 97%
Purity: 97%
CAS No.: 17920-35-3
Molecular Formula: C6H8N2O
Formula Weight: 124.14
Physical Form: Liquid
Sensitivity: Air Sensitive
Appearance: Brown
Beilstein Reference: 115179
UN No.: 2810
MDL No.: MFCD04972542
Formula:
C₆H₈N₂O MW: 124.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04972542 CAS Number: 17920-35-3 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...