(2-Aminophenyl)acetonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
2-Aminophenylacetonitrile, 97%
Purity: 97%
CAS No.: 2973-50-4
Molecular Formula: C8H8N2
Formula Weight: 132.17
Physical Form: Powder
Appearance: Off-white
Melting Point: 69-72°
Beilstein Reference: 2716483
UN No.: 3439
MDL No.: MFCD00091082
Formula:
C₈H₈N₂ Melting Pt: 69…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00091082 CAS Number: 2973-50-4 EINECS: 221-015-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...