1-bromo-2,4-dimethyl-5-nitrobenzene 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-2,4-dimethyl-5-nitrobenzene
H66208-06
H66208-14
AAH66208-03EA
333
USD
AAH66208-03
AAH66208-06
AAH66208-14
1-bromo-2,4-dimethyl-5-nitrobenzene 95%
1-bromo-2,4-dimethyl-5-nitrobenzene
1-Bromo-2,4-dimethyl-5-nitrobenzene, 95%
Purity: 95%
CAS No.: 69383-59-1
Molecular Formula: C8H8BrNO2
Formula Weight: 230.06
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Pale yellow
Beilstein Reference: 1948547
UN No.: 2811
MDL No.: MFCD00075824
Formula:
C₈H₈BrNO₂ MW: 230.061 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00075824 CAS Number: 69383-59-1 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...