2-Aminonicotinamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Aminonicotinamide, 2-Aminopyridine-3-carboxamide
H66202-03
H66202-06
AAH66202-03EA
59.3
USD
AAH66202-03
AAH66202-06
2-Aminonicotinamide 97%
2-Aminonicotinamide
2-Aminonicotinamide, 97%
Purity: 97%
CAS No.: 13438-65-8
Molecular Formula: C6H7N3O
Formula Weight: 137.14
Physical Form: Solid
Appearance: Pale yellow
Melting Point: 194-200°
Beilstein Reference: 3613
UN No.: 2811
MDL No.: MFCD00128873
Formula:
C₆H₇N₃O MW: 137.142 g/mol Melting Pt: 194…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128873 CAS Number: 13438-65-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...